missing translation for 'onlineSavingsMsg'
Learn More

1,3-Adamantanediol, TRC

Product Code. 30135262
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135262 25 g 25g
30132963 10 g 10g
30010007 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135262 Supplier Toronto Research Chemicals Supplier No. TRCA20808025G
 Restricted Item
Estimated Shipment: 22-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5001-18-3
Molecular Formula C10H16O2
Molecular Weight (g/mol) 168.23
Synonym 1,3-Adamantandiol, 1,3-Dihydroxyadamantane;
IUPAC Name adamantane-1,3-diol
SMILES O[C@@]1(C[C@]2(O)C3)C[C@H](C2)C[C@H]3C1

Specifications

Quantity 25 g
InChI Formula InChI=1S/C10H16O2/c11-9-2-7-1-8(4-9)5-10(12,3-7)6-9/h7-8,11-12H,1-6H2
Formula Weight 168.115
Chemical Name or Material 1,3-Adamantanediol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.