missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Adamantanedicarboxylic acid, TRC
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Specifications
Specifications
| Chemical Name or Material | 1,3-Adamantanedicarboxylic acid |
| CAS | 39269-10-8 |
| Molecular Formula | C12H16O4 |
| Quantity | 250 mg |
| Synonym | 1,3-Dicarboxyadamantane |
| InChI Formula | InChI=1S/C12H16O4/c13-9(14)11-2-7-1-8(4-11)5-12(3-7,6-11)10(15)16/h7-8H,1-6H2,(H,13,14)(H,15,16) |
| SMILES | OC(=O)C12CC3CC(CC(C3)(C1)C(=O)O)C2 |
| IUPAC Name | adamantane-1,3-dicarboxylic acid |
| Molecular Weight (g/mol) | 224.25 |
| Formula Weight | 224.1049 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction