missing translation for 'onlineSavingsMsg'
Learn More

1,3,6,8-Pyrenetetrasulfonic acid tetrasodium salt, 95%

Product Code. p-189180 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10695721

Brand: Thermo Scientific Acros 325560010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 59572-10-0 | C16H6Na4O12S4 | 610.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59572-10-0
Molecular Formula C16H6Na4O12S4
Molecular Weight (g/mol) 610.44
MDL Number MFCD00042029
InChI Key UZBIRLJMURQVMX-UHFFFAOYSA-J
Synonym 1,3,6,8-pyrenetetrasulfonic acid, tetrasodium salt, unii-p062ld4v71, 1,3,6,8-pyrenetetrasulfonic acid tetrasodium salt, tetrasodium 1,3,6,8-pyrenetetrasulfonate, tetrasodium pyrene-1,3,6,8-tetrasulfonate, 1,3,6,8-pyrenetetrasulfonic acid, sodium salt 1:4, 1,3,6,8-pyrenetetrasulfonic acid tetrasodium salt hydrate, zlchem 1090, dsstox_cid_21233, dsstox_gsid_41233
PubChem CID 101082
IUPAC Name tetrasodium;pyrene-1,3,6,8-tetrasulfonate
SMILES C1=CC2=C3C(=C(C=C2S(=O)(=O)[O-])S(=O)(=O)[O-])C=CC4=C(C=C(C1=C43)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+].[Na+].[Na+]

Specifications

CAS Min % 80.0
CAS Max % 100.0
Color Green
Infrared Spectrum Authentic
Assay Percent Range 85%
Packaging Glass bottle
Quantity 1 g
Formula Weight 610.44
Percent Purity 85%
Physical Form Fine Crystalline Powder
Chemical Name or Material 1,3,6,8-Pyrenetetrasulfonic acid tetrasodium salt, 85%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.