missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Tris(2-hydroxyethyl)cyanuric acid, 97%

Product Code. 11339739
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11339739 50 g 50g
11349739 250 g 250g
11329739 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11339739 Supplier Thermo Scientific Alfa Aesar Supplier No. B21697.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 839-90-7 | C9H15N3O6 | 261.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 839-90-7
Molecular Formula C9H15N3O6
Molecular Weight (g/mol) 261.23
MDL Number MFCD00003549
InChI Key BPXVHIRIPLPOPT-UHFFFAOYSA-N
Synonym 1,3,5-tris 2-hydroxyethyl cyanuric acid, theic, tris 2-hydroxyethyl isocyanurate, tris hydroxyethyl cyanurate, tris 2-hydroxyethyl cyanurate, tris hydroxyethyl isocyanurate, 1,3,5-triazine-2,4,6 1h,3h,5h-trione, 1,3,5-tris 2-hydroxyethyl, 1,3,5-tris 2-hydroxyethyl-1,3,5-triazinane-2,4,6-trione, unii-n7w2mh0p2m
PubChem CID 13286
SMILES OCCN1C(=O)N(CCO)C(=O)N(CCO)C1=O

Specifications

Melting Point 136°C to 140°C
Quantity 50 g
IUPAC Name tris(2-hydroxyethyl)-1,3,5-triazinane-2,4,6-trione
Formula Weight 261.24
Percent Purity 97%
Chemical Name or Material 1,3,5-Tris(2-hydroxyethyl)cyanuric acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.