missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Trioxane, TRC

Product Code. 30449956
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30449956 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30449956 Supplier Toronto Research Chemicals Supplier No. TRCT805550250G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,3,5-Trioxane
CAS 110-88-3
Molecular Formula C3 H6 O3
Quantity 250 g
Synonym 1,3,5-Trioxane, Trioxane (6CI,7CI), s-Trioxane (8CI), Formaldehyde','trimer, NSC 26347, T 0505, Triformol, Trioxan, Trioxymethylene, sym-Trioxane
InChI Formula InChI=1S/C3H6O3/c1-4-2-6-3-5-1/h1-3H2
SMILES C1OCOCO1
IUPAC Name 1,3,5-trioxane
Molecular Weight (g/mol) 90.08
Formula Weight 90.0317
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.