missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Trimethyl-1H-pyrazole-4-carboxylic Acid, TRC

Product Code. 30345273
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30345273 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30345273 Supplier Toronto Research Chemicals Supplier No. TRCB535268100MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Chemical Name or Material 1,3,5-Trimethyl-1H-pyrazole-4-carboxylic Acid
CAS 1125-29-7
Molecular Formula C7 H10 N2 O2
Quantity 100 mg
InChI Formula InChI=1S/C7H10N2O2/c1-4-6(7(10)11)5(2)9(3)8-4/h1-3H3,(H,10,11)
SMILES Cc1nn(C)c(C)c1C(=O)O
IUPAC Name 1,3,5-trimethylpyrazole-4-carboxylic acid
Molecular Weight (g/mol) 154.167
Formula Weight 154.074
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.