missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Triisopropylbenzene, 95%

Product Code. 11435523
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11435523 50 g 50g
11445523 250 g 250g
11425523 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11435523 Supplier Thermo Scientific Alfa Aesar Supplier No. A13453.18
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 717-74-8 | C15H24 | 204.357 g/mol

1,3,5-Triisopropylbenzene is used as a fuel and fuel additive. It is also used in lubricants and lubricant additives. Further, it is used in the preparation of 2,4,6-triisopropyl-benzenesulfonyl chloride. It is also employed as a swelling agent during the synthesis of mesoporous silica with two-dimensional hexagonal pores. In addition to this, it is used as a micelle expander in the synthesis of magnetic mesoporous silica nanoparticles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3,5-Triisopropylbenzene is used as a fuel and fuel additive. It is also used in lubricants and lubricant additives. Further, it is used in the preparation of 2,4,6-triisopropyl-benzenesulfonyl chloride. It is also employed as a swelling agent during the synthesis of mesoporous silica with two-dimensional hexagonal pores. In addition to this, it is used as a micelle expander in the synthesis of magnetic mesoporous silica nanoparticles.

Solubility
Immiscible with water.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 717-74-8
Molecular Formula C15H24
Molecular Weight (g/mol) 204.357
MDL Number MFCD00008890
InChI Key VUMCUSHVMYIRMB-UHFFFAOYSA-N
Synonym 1,3,5-triisopropylbenzene, 2,4,6-triisopropylbenzene, benzene, 1,3,5-tris 1-methylethyl, 1,3,5-tris propan-2-yl benzene, unii-fr9y346wpb, benzene, 1,3,5-triisopropyl, fr9y346wpb, benzene, tris 1-methylethyl, pubchem13720, 1,5-triisopropylbenzene
PubChem CID 12860
IUPAC Name 1,3,5-tri(propan-2-yl)benzene
SMILES CC(C)C1=CC(=CC(=C1)C(C)C)C(C)C

Specifications

Melting Point -7°C
Density 0.854
Boiling Point 235°C to 237°C
Flash Point 86°C (186°F)
Refractive Index 1.488
Quantity 50 g
Beilstein 1862750
Solubility Information Immiscible with water.
Formula Weight 204.36
Percent Purity 95%
Chemical Name or Material 1,3,5-Triisopropylbenzene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.