missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Trichloro-2-nitrobenzene, 98+%

Product Code. 11414973
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11414973 25 g 25g
11404973 5 g 5g
11424973 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11414973 Supplier Thermo Scientific Alfa Aesar Supplier No. A12262.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 18708-70-8 | C6H2Cl3NO2 | 226.437 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18708-70-8
Molecular Formula C6H2Cl3NO2
Molecular Weight (g/mol) 226.437
MDL Number MFCD00014690
InChI Key AEBJDOTVYMITIA-UHFFFAOYSA-N
PubChem CID 29228
IUPAC Name 1,3,5-trichloro-2-nitrobenzene
SMILES C1=C(C=C(C(=C1Cl)[N+](=O)[O-])Cl)Cl

Specifications

Melting Point 68°C to 73°C
Quantity 25 g
UN Number UN2811
Beilstein 2213282
Synonym 2,4,6-trichloronitrobenzene, benzene, 1,3,5-trichloro-2-nitro, 2,4,6-trichloro-1-nitrobenzene, unii-t00kp6ct7k, t00kp6ct7k, benzene,1,3,5-trichloro-2-nitro, acmc-209epa, 2,6-trichloronitrobenzene, dsstox_cid_6204, dsstox_rid_78056
Formula Weight 226.45
Percent Purity ≥98%
Chemical Name or Material 1,3,5-Trichloro-2-nitrobenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.