missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Trichloro-2,4,6-trifluorobenzene, 98%, Thermo Scientific™

Catalog Number 11492888
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
319-88-0
C6Cl3F3
235.411
MFCD00000587
QPXZZPSKCVNHFW-UHFFFAOYSA-N
1,3,5-trichlorotrifluorobenzene, benzene, 1,3,5-trichloro-2,4,6-trifluoro, sym-trichlorotrifluorobenzene, sym-trifluorotrichlorobenzene, 1,3,5-trifluorotrichlorobenzene, 2,4,6-trifluorotrichlorobenzene, 1,3,5-trifluoro-2,4,6-trichlorobenzene, acmc-20ap5m, 1,5-trichlorotrifluorobenzene, wln: gr cg eg bf df ff
67573
1,3,5-trichloro-2,4,6-trifluorobenzene
C1(=C(C(=C(C(=C1Cl)F)Cl)F)Cl)F
This item is not returnable. View return policy
319-88-0
C6Cl3F3
235.411
MFCD00000587
QPXZZPSKCVNHFW-UHFFFAOYSA-N
1,3,5-trichlorotrifluorobenzene, benzene, 1,3,5-trichloro-2,4,6-trifluoro, sym-trichlorotrifluorobenzene, sym-trifluorotrichlorobenzene, 1,3,5-trifluorotrichlorobenzene, 2,4,6-trifluorotrichlorobenzene, 1,3,5-trifluoro-2,4,6-trichlorobenzene, acmc-20ap5m, 1,5-trichlorotrifluorobenzene, wln: gr cg eg bf df ff
67573
1,3,5-trichloro-2,4,6-trifluorobenzene
C1(=C(C(=C(C(=C1Cl)F)Cl)F)Cl)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 62°C to 64°C
Boiling Point 79°C to 80°C (12 mmHg)
Flash Point 102°C (215°F)
Quantity 5 g
Beilstein 958307
Formula Weight 235.42
Percent Purity 98%
Chemical Name or Material 1,3,5-Trichloro-2,4,6-trifluorobenzene
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'