missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Tribromobenzene, 98%

Product Code. 11466706
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11466706 500 g 500g
11446706 25 g 25g
11456706 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11466706 Supplier Thermo Scientific Alfa Aesar Supplier No. B23860.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 626-39-1 | C6H3Br3 | 314.80 g/mol

1,3,5-Tribromobenzene is used as an intermediate for active pharmaceutical ingredient manufacturing. It acts as an intermediate for the preparation of 1,3,5-trimethoxytoluene. It forms molecular complexes with [60]-and [70] fullerenes. Further, it is employed as an internal standard during derivatization step in the determination of cyanide in human plasma and urine by gas chromatography-mass spectrometry. In addition to this, it reacts with the perfluoroalkenylzinc reagent in the presence of tetrakis(triphenylphosphine)palladium catalyst to yield a trifunctional monomer 1,3,5-tris(alfa, beta, beta-trifluorovinyl)benzene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3,5-Tribromobenzene is used as an intermediate for active pharmaceutical ingredient manufacturing. It acts as an intermediate for the preparation of 1,3,5-trimethoxytoluene. It forms molecular complexes with [60]-and [70] fullerenes. Further, it is employed as an internal standard during derivatization step in the determination of cyanide in human plasma and urine by gas chromatography-mass spectrometry. In addition to this, it reacts with the perfluoroalkenylzinc reagent in the presence of tetrakis(triphenylphosphine)palladium catalyst to yield a trifunctional monomer 1,3,5-tris(alfa, beta, beta-trifluorovinyl)benzene.

Solubility
Soluble in hot ethanol and acetic acid. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 626-39-1
Molecular Formula C6H3Br3
Molecular Weight (g/mol) 314.80
MDL Number MFCD00000080
InChI Key YWDUZLFWHVQCHY-UHFFFAOYSA-N
Synonym benzene, 1,3,5-tribromo, 1,3,5-tribromo-benzene, unii-o3td0u1oaq, o3td0u1oaq, paragos 530416, 1,5-tribromobenzene, pubchem9590, 1,3,5-tribromobezene, benzene,3,5-tribromo, acmc-20a11o
PubChem CID 12279
IUPAC Name 1,3,5-tribromobenzene
SMILES BrC1=CC(Br)=CC(Br)=C1

Specifications

Melting Point 121°C to 124°C
Boiling Point 271°C
Quantity 500 g
Beilstein 1858917
Solubility Information Soluble in hot ethanol and acetic acid. Insoluble in water.
Formula Weight 314.8
Percent Purity 98%
Chemical Name or Material 1,3,5-Tribromobenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.