missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Triallyl-1,3,5-triazinane-2,4,6-trione, TRC

Product Code. 30450690
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
Unit Size:
2.50g
500mg
Product Code. Quantity unitSize
30450690 250 mg -
30451539 2.5 g 2.50g
30456048 500 mg 500mg
3 options
This item is not returnable. View return policy

Product Code. 30450690

Brand: Toronto Research Chemicals TRCT767153250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1025-15-6
Formula Weight 249.1113
InChI Formula InChI=1S/C12H15N3O3/c1-4-7-13-10(16)14(8-5-2)12(18)15(9-6-3)11(13)17/h4-6H,1-3,7-9H2
IUPAC Name 1,3,5-tris(prop-2-enyl)-1,3,5-triazinane-2,4,6-trione
Molecular Formula C12 H15 N3 O3
Molecular Weight (g/mol) 249.2658
SMILES C=CCN1C(=O)N(CC=C)C(=O)N(CC=C)C1=O
Synonym 1,3,5-Triazine-2,4,6(1H,3H,5H)-trione, 1,3,5-tri-2-propen-1-yl-, 1,3,5-Triazine-2,4,6(1H,3H,5H)-trione, 1,3,5-tri-2-propenyl- (9CI), s-Triazine-2,4,6(1H,3H,5H)-trione, 1,3,5-triallyl- (8CI), s-Triazine-2,4,6(1H,3H,5H)-trione, triallyl- (6CI,7CI), 1,3,5-Tri-2-propen-1-yl-1,3,5-triazine-2,4,6(1H,3H,5H)-trione, 1,3,5-Tri-2-propenyl-1,3,5-triazine-2,4,6(1H,3H,5H)-trione, 1,3,5-Triallyl-1,3,5-triazin-2,4,6-trione, 1,3,5-Triallyl-1,3,5-triazine-2,4,6(1H,3H,5H)-trione, 1,3,5-Triallylisocyanurate, 1,3,5-Triallylisocyanuric acid, 1,3,5-Tris(2-propenyl) isocyanurate, 1,3,5-Tris-2'-propenylisocyanuric acid, Betalink IC/W 65PA6, CVD 50106, DLC-A, Daik 7, Diak 7, Drimix TAIC, Drimix TAIC 75, HBSQ 2011, Isocyanuric acid triallyl ester, Kettlitz TAIC, Luvomaxx TAIC-DL 70, Luvomaxx TAIC-DL 70C, M 6, M 6 (isocyanurate), M 60, M 60 (isocyanurate), NSC 11692, Nor-TAIC KS, PL 301, PL 301-50D, Perkalink 301, Perkalink 301-50, Perkalink 301-501, Perkalink 301-50D, Perkalink 301-70DPD, Rhenofit TAIC/S, SR 533, SR 533R, Synpro PLC 4185, T 123406, TAIC, TAIC 100, TAIC 60, TAIC 70, TAIC DLC-A, TAIC WH, TAIC-B, TAIC-M 6, TAIC-M 60, TAIC-S, TAIC-WH 60, Taic-S 70, Taicros, Triallyl isocyanurate, Triallyl-1,3,5-triazine-2,4,6(1H,3H,5H)-trione, Triallyl-1,3,5-triazine-2,4,6-trione, Triallylisocyanuric acid, Trilink 7, WH 60
Chemical Name or Material Triallyl isocyanurate

Specifications

Quantity 250 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.