missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3,5-Tri-O-benzoyl-alpha-D-ribofuranose, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
It is employed as a starting material for nucleosides and is an intermediate fir cloforabine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is employed as a starting material for nucleosides and is an intermediate fir cloforabine.
Solubility
Insoluble in water, easily soluble in chloroform.
Notes
Store at room temperature. Keep away from oxidizing agents and heats.
It is employed as a starting material for nucleosides and is an intermediate fir cloforabine.
Solubility
Insoluble in water, easily soluble in chloroform.
Notes
Store at room temperature. Keep away from oxidizing agents and heats.
Specifications
Melting Point | 125°C to 129°C |
Color | Brown |
Quantity | 1 g |
Beilstein | 98446 |
Solubility Information | Insoluble in water,easily soluble in chloroform. |
Formula Weight | 462.45 |
Percent Purity | 97% |
Physical Form | Powder |
Chemical Name or Material | 1,3,5-Tri-O-benzoyl-alpha-D-ribofuranose |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'