missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 1,3,5-Tri-O-benzoyl-2-deoxy-2-fluoro-alpha-D-arabinofuranose

Product Code. 15400657
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15400657

Brand: Thermo Scientific Chemicals J66805.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 97614-43-2 | C26H21FO7 | 464.445 g/mol

It is a pharmaceutical intermediate and is used as an OLED materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a pharmaceutical intermediate and is used as an OLED materials.

Solubility
Soluble in chloroform or dichloromethane

Notes
Refrigerate. Keep away from heat, oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97614-43-2
Molecular Formula C26H21FO7
Molecular Weight (g/mol) 464.445
MDL Number MFCD00083339
InChI Key JOAHVPNLVYCSAN-UXGLMHHASA-N
Synonym 2-deoxy-2-fluoro-1,3,5-tri-o-benzoyl-alpha-d-arabinofuranose, 2r,3s,4r,5r-5-benzoyloxy methyl-3-fluorotetrahydrofuran-2,4-diyl dibenzoate, 2-deoxy-2-fluoro-1,3,5-tri-o-benzoyl-, a-d-arabinofuranose, 1,3,5-tri-o-benzoyl-2-deoxy-2-fluoro-a-d-arabinofuranose, 2r,3r,4s,5r-3,5-bis benzoyloxy-4-fluorooxolan-2-yl methyl benzoate, 2r,3s,4r,5r-5-benzoyloxymethyl-3-fluoro-tetrahydrofuran-2,4-diyl dibenzoate, 1,3,5-tri-o-benzoyl-2-deoxy-2-fluoro-md-arabinose, 2-deoxy-2-fluoro-1,3,5-tri-o-benzoyl-d-ribofuranose, pubchem20401, 2-deoxy-2-fluoro-1,3,5-tri-o-benzoyl-alpha-d-ribofuranose
PubChem CID 11754171
IUPAC Name [(2R,3R,4S,5R)-3,5-dibenzoyloxy-4-fluorooxolan-2-yl]methyl benzoate
SMILES C1=CC=C(C=C1)C(=O)OCC2C(C(C(O2)OC(=O)C3=CC=CC=C3)F)OC(=O)C4=CC=CC=C4

Specifications

Melting Point 76°C to 78°C
Color White
Quantity 5 g
Solubility Information Soluble in chloroform or dichloromethane
Formula Weight 464.44
Physical Form Powder
Chemical Name or Material 1,3,5-Tri-O-benzoyl-2-deoxy-2-fluoro-alpha-D-arabinofuranose

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.