missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Tri(anthracen-9-yl)benzene (contains ~20% inorganics), TRC

Product Code. 30454705
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30454705 50 mg 50mg
30449851 100 mg 100mg
30451260 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30454705 Supplier Toronto Research Chemicals Supplier No. TRCT89676350MG
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 713542-04-2
Molecular Formula C48H30
Molecular Weight (g/mol) 606.75
Synonym 9,9',9' '-(1,3,5-benzenetriyl)tris-Anthracene, 9,9’,9’’-(1,3,5-Benzenetriyl)tris[anthracene], 1,3,5-Tri(9-anthryl)benzene
IUPAC Name 9-[3,5-di(anthracen-9-yl)phenyl]anthracene
SMILES c1ccc2c(c3cc(cc(c3)c4c5ccccc5cc6ccccc46)c7c8ccccc8cc9ccccc79)c%10ccccc%10cc2c1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C48H30/c1-7-19-40-31(13-1)25-32-14-2-8-20-41(32)46(40)37-28-38(47-42-21-9-3-15-33(42)26-34-16-4-10-22-43(34)47)30-39(29-37)48-44-23-11-5-17-35(44)27-36-18-6-12-24-45(36)48/h1-30H
Formula Weight 606.2348
Chemical Name or Material 1,3,5-Tri(anthracen-9-yl)benzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.