missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Tri(4-carboxyphenyl)benzene, 97%

Product Code. 15446558
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15446558 5 g 5g
15436558 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15446558 Supplier Thermo Scientific Alfa Aesar Supplier No. H60087.06
 Restricted Item
Estimated Shipment: 13-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 50446-44-1 | C27H18O6 | 438.44 g/mol

1,3,5-Tri(4-carboxyphenyl)benzene is used as a building block for metal organic frameworks, which is a 3D-microporous material and finds applications in gas adsorption and separation technologies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3,5-Tri(4-carboxyphenyl)benzene is used as a building block for metal organic frameworks, which is a 3D-microporous material and finds applications in gas adsorption and separation technologies.

Solubility
Soluble in tetrahydrofuran.

Notes
Incompatible with strong oxidizing agents, metals, amines, strong acids, acid chlorides, alcohols, peroxides, permanganates, strong reducing agents, soluble carbonates, phosphates, phosphorus halides and hydroxides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50446-44-1
Molecular Formula C27H18O6
Molecular Weight (g/mol) 438.44
MDL Number MFCD10000888
InChI Key SATWKVZGMWCXOJ-UHFFFAOYSA-N
Synonym 1,3,5-tris 4-carboxyphenyl benzene, 1,3,5-tri 4-carboxyphenyl benzene, 5'-4-carboxyphenyl-1,1':3',1-terphenyl-4,4-dicarboxylic acid, 1,1':3',1-terphenyl-4,4-dicarboxylic acid, 5'-4-carboxyphenyl, acmc-209kmn, h3btb, 4,4?,4??,-benzene-1,3,5-triyl-tris benzoic acid, 1,3,5-tris 4-carboxyphenyl benzene, ≤20 wt. % solvent, 3',5'-bis 4-carboxyphenyl-1,1'-biphenyl-4-carboxylic acid, 5'-4-carboxyphenyl-1,1':3',1-terphenyl-4,4-dicarboxylicacid
PubChem CID 10694305
IUPAC Name 4-[3,5-bis(4-carboxyphenyl)phenyl]benzoic acid
SMILES OC(=O)C1=CC=C(C=C1)C1=CC(=CC(=C1)C1=CC=C(C=C1)C(O)=O)C1=CC=C(C=C1)C(O)=O

Specifications

Melting Point 322°C to 327°C
Quantity 5 g
UN Number UN3077
Solubility Information Soluble in tetrahydrofuran.
Formula Weight 438.43
Percent Purity 97%
Chemical Name or Material 1,3,5-Tri(4-carboxyphenyl)benzene
compliance-icons compliance-icons
Product Identifier
  • 1,3,5-Tri(4-carboxyphenyl)benzene
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 1,3,5-Tris(4-carboxyphenyl)benzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.