missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Pentanetriol, TRC

Product Code. 30413857
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
250mg
Product Code. Quantity unitSize
30413857 2.5 g 2.5g
30420870 250 mg 250mg
2 options
This item is not returnable. View return policy

Product Code. 30413857

Brand: Toronto Research Chemicals TRCP2740452.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4328-94-3
Formula Weight 120.0786
InChI Formula InChI=1S/C5H12O3/c6-3-1-5(8)2-4-7/h5-8H,1-4H2
IUPAC Name pentane-1,3,5-triol
Molecular Formula C5H12O3
Molecular Weight (g/mol) 120.15
SMILES OCCC(O)CCO
Chemical Name or Material 1,3,5-Pentanetriol

Specifications

Quantity 2.5 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.