missing translation for 'onlineSavingsMsg'
Learn More

1-(3,5-Dichlorophenyl)-5-propyl-1H-pyrazole-4-carboxylic acid, 97%, Thermo Scientific™

Catalog Number 10273712
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
306936-60-7
C13H12Cl2N2O2
299.151
MFCD02677727
CIAUICOJDLJNBL-UHFFFAOYSA-N
1-3,5-dichlorophenyl-5-propyl-1h-pyrazole-4-carboxylic acid, 1-3,5-dichlorophenyl-5-propylpyrazole-4-carboxylic acid, 1h-pyrazole-4-carboxylicacid, 1-3,5-dichlorophenyl-5-propyl, maybridge3_006729, 4-carboxy-1-3,5-dichlorophenyl-5-propyl-1h-pyrazole
2743017
1-(3,5-dichlorophenyl)-5-propylpyrazole-4-carboxylic acid
CCCC1=C(C=NN1C2=CC(=CC(=C2)Cl)Cl)C(=O)O
This item is not returnable. View return policy
306936-60-7
C13H12Cl2N2O2
299.151
MFCD02677727
CIAUICOJDLJNBL-UHFFFAOYSA-N
1-3,5-dichlorophenyl-5-propyl-1h-pyrazole-4-carboxylic acid, 1-3,5-dichlorophenyl-5-propylpyrazole-4-carboxylic acid, 1h-pyrazole-4-carboxylicacid, 1-3,5-dichlorophenyl-5-propyl, maybridge3_006729, 4-carboxy-1-3,5-dichlorophenyl-5-propyl-1h-pyrazole
2743017
1-(3,5-dichlorophenyl)-5-propylpyrazole-4-carboxylic acid
CCCC1=C(C=NN1C2=CC(=CC(=C2)Cl)Cl)C(=O)O
CAS Min % 97.0
Assay Percent Range 97%
Quantity 1 g
Formula Weight 299.16
Percent Purity 97%
Chemical Name or Material 1-(3, 5-Dichlorophenyl)-5-propyl-1H-pyrazole-4-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'