missing translation for 'onlineSavingsMsg'
Learn More

1-(3,5-Dichloro-4-pyridyl)piperazine, ≥95%, Thermo Scientific™

Product Code. 10487442
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 10487442

Brand: Thermo Scientific Maybridge SPB06834DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 175277-80-2
Molecular Formula C9H11Cl2N3
Molecular Weight (g/mol) 232.11
MDL Number MFCD00221400
InChI Key GPCFTLKHUONISF-UHFFFAOYSA-N
Synonym 1-3,5-dichloropyridin-4-yl piperazine, 1-3,5-dichloro-4-pyridyl piperazine, piperazine, 1-3,5-dichloro-4-pyridinyl, buttpark 4357-83, 3,5-dichloro-4-pyridyl piperazine, 1-3,5-dichloro-4-pyridinyl piperazine, 1-3,5-dichloro-pyridin-4-yl-piperazine, piperazine,1-3,5-dichloro-4-pyridinyl, 1-3,5-bis chloranyl pyridin-4-yl piperazine
PubChem CID 2799984
IUPAC Name 1-(3,5-dichloropyridin-4-yl)piperazine
SMILES ClC1=CN=CC(Cl)=C1N1CCNCC1

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 232.11
Percent Purity 95%
Chemical Name or Material 1-(3, 5-Dichloro-4-pyridyl)piperazine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.