missing translation for 'onlineSavingsMsg'
Learn More

1-(3,5-Di-tert-butyl-4-hydroxyphenyl)ethanone, TRC

Product Code. 30341152
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 30341152

Brand: Toronto Research Chemicals TRCB415748500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3,5-Di-tert-butyl-4-hydroxyacetophenone
CAS 14035-33-7
Molecular Formula C16 H24 O2
Quantity 500 mg
Synonym Acetophenone, 3',5'-di-tert-butyl-4'-hydroxy- (6CI,7CI,8CI), 1-[3,5-Bis(1,1-dimethylethyl)-4-hydroxyphenyl]ethanone, 1-(3,5-Ditert-butyl-4-hydroxyphenyl)ethanone, 1-[3,5-Di(tert-butyl)-4-hydroxyphenyl]ethan-1-one, 2,6-Di-tert-butyl-4-acetylphenol, 3,5-Di-tert-butyl-4-hydroxyphenyl methyl ketone, 3',5'-Di-tert-butyl-4'-hydroxyacetophenone, 4-Acetyl-2,6-di-tert-butylphenol, 4'-Hydroxy-3',5'-di-tert-butylacetophenone, 3,5-Di-tert-butyl-4-hydroxyacetophenone
InChI Formula InChI=1S/C16H24O2/c1-10(17)11-8-12(15(2,3)4)14(18)13(9-11)16(5,6)7/h8-9,18H,1-7H3
SMILES CC(=O)c1cc(c(O)c(c1)C(C)(C)C)C(C)(C)C
IUPAC Name 1-(3,5-ditert-butyl-4-hydroxyphenyl)ethanone
Molecular Weight (g/mol) 248.3606
Formula Weight 248.1776
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.