missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(3,5-Di-tert-butyl-4-hydroxyphenyl)ethanone, TRC
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Specifications
Specifications
| Chemical Name or Material | 3,5-Di-tert-butyl-4-hydroxyacetophenone |
| CAS | 14035-33-7 |
| Molecular Formula | C16 H24 O2 |
| Quantity | 50 mg |
| Synonym | Acetophenone, 3',5'-di-tert-butyl-4'-hydroxy- (6CI,7CI,8CI), 1-[3,5-Bis(1,1-dimethylethyl)-4-hydroxyphenyl]ethanone, 1-(3,5-Ditert-butyl-4-hydroxyphenyl)ethanone, 1-[3,5-Di(tert-butyl)-4-hydroxyphenyl]ethan-1-one, 2,6-Di-tert-butyl-4-acetylphenol, 3,5-Di-tert-butyl-4-hydroxyphenyl methyl ketone, 3',5'-Di-tert-butyl-4'-hydroxyacetophenone, 4-Acetyl-2,6-di-tert-butylphenol, 4'-Hydroxy-3',5'-di-tert-butylacetophenone, 3,5-Di-tert-butyl-4-hydroxyacetophenone |
| InChI Formula | InChI=1S/C16H24O2/c1-10(17)11-8-12(15(2,3)4)14(18)13(9-11)16(5,6)7/h8-9,18H,1-7H3 |
| SMILES | CC(=O)c1cc(c(O)c(c1)C(C)(C)C)C(C)(C)C |
| IUPAC Name | 1-(3,5-ditert-butyl-4-hydroxyphenyl)ethanone |
| Molecular Weight (g/mol) | 248.3606 |
| Formula Weight | 248.1776 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction