missing translation for 'onlineSavingsMsg'
Learn More

1-[3-(4-Methylpiperazin-1-yl)phenyl]methanamine, 97%, Thermo Scientific™

Product Code. 10193324
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10193324 5 g Amber Glass Bottle 5g
10351053 250 mg Amber Glass Bottle 250mg
10695713 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10193324 Supplier Thermo Scientific Maybridge Supplier No. CC35913DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 672325-37-0
Molecular Formula C12H19N3
Molecular Weight (g/mol) 205.305
InChI Key VKTLTRIKVYJVOX-UHFFFAOYSA-N
Synonym 1-3-4-methylpiperazin-1-yl phenyl methanamine, 3-4-methyl-1-piperazinyl benzenemethanamine, 3-4-methylpiperazin-1-yl phenyl methanamine, 3-4-methyl-1-piperazinyl-benzenemethanamine, 3-4-methyl-piperazin-1-yl-benzylamine, 3-4-methyl-1-piperazinyl-benzene methanamine, 3-4-methylpiperazinyl phenyl methylamine, pubchem12998
PubChem CID 4961252
IUPAC Name [3-(4-methylpiperazin-1-yl)phenyl]methanamine
SMILES CN1CCN(CC1)C2=CC=CC(=C2)CN

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 205.3
Percent Purity 97%
Chemical Name or Material 1-[3-(4-Methylpiperazin-1-yl)phenyl]methanamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.