missing translation for 'onlineSavingsMsg'
Learn More

1-(3,4-Dimethoxyphenyl)propan-2-one, Mikromol™

Product Code. 17960045
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17960045 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17960045 Supplier Mikromol Supplier No. MM1501.140100
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

1-(3,4-Dimethoxyphenyl)propan-2-one
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-(3,4-Dimethoxyphenyl)propan-2-one
CAS 776-99-8
Analyte or Component Names 1-(3,4-Dimethoxyphenyl)propan-2-one
Shipping Condition Room Temperature
Molecular Formula C11 H14 O3
Certifications/Compliance ISO 9001
Synonym 2-Propanone, 1-(3,4-dimethoxyphenyl)-, 2-Propanone, (3,4-dimethoxyphenyl)- (6CI,7CI), 1-(3,4-Dimethoxyphenyl)-2-propanone, (3,4-Dimethoxyphenyl)acetone, 1-(3,4-Dimethoxyphenyl)acetone, 3,4-Dimethoxybenzyl methyl ketone, NSC 16700, Veratryl acetone
InChI Formula InChI=1S/C11H14O3/c1-8(12)6-9-4-5-10(13-2)11(7-9)14-3/h4-5,7H,6H2,1-3H3
SMILES COc1ccc(CC(=O)C)cc1OC
IUPAC Name 1-(3,4-dimethoxyphenyl)propan-2-one
Molecular Weight (g/mol) 194.23
Quantity 100 mg
Formula Weight 194.0943 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.