missing translation for 'onlineSavingsMsg'
Learn More

1-(3',4'-Dimethoxyphenyl)-1-propanol, TRC

Product Code. 30372145
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30372145 500 mg
30380249 1 g
30374845 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30372145 Supplier Toronto Research Chemicals Supplier No. TRCD470740500MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10548-83-1
Molecular Formula C11H16O3
Molecular Weight (g/mol) 196.24
Synonym alpha-Ethyl-3,4-dimethoxybenzyl Alcohol, alpha-Ethylveratryl Alcohol, Ethylveratrylcarbinol, 1-Veratryl Propanol
SMILES CCC(O)c1ccc(OC)c(OC)c1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C11H16O3/c1-4-9(12)8-5-6-10(13-2)11(7-8)14-3/h5-7,9,12H,4H2,1-3H3
Formula Weight 196.1099
Chemical Name or Material 1-(3',4'-Dimethoxyphenyl)-1-propanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.