missing translation for 'onlineSavingsMsg'
Learn More

1-(3,4-Dihydroxyphenyl)-2-[(phenylmethyl)amino]-ethanone-d5, TRC

Product Code. 30385797
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
10 mg
Unit Size:
10mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30385797 5 mg 5mg
30378275 10 mg 10mg
30373652 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30385797 Supplier Toronto Research Chemicals Supplier No. TRCD4543675MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-(3,4-Dihydroxyphenyl)-2-[(phenylmethyl)amino]-ethanone-d5
Molecular Formula C15 D5 H10 N O3
Quantity 5 mg
Synonym NSC 33301-d5
InChI Formula InChI=1S/C15H15NO3/c17-13-7-6-12(8-14(13)18)15(19)10-16-9-11-4-2-1-3-5-11/h1-8,16-18H,9-10H2/i6D,7D,8D,10D2
SMILES [2H]c1c([2H])c(C(=O)C([2H])([2H])NCc2ccccc2)c([2H])c(O)c1O
IUPAC Name 2-(benzylamino)-2,2-dideuterio-1-(2,3,6-trideuterio-4,5-dihydroxyphenyl)ethanone
Molecular Weight (g/mol) 262.315
Formula Weight 262.137
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.