missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 1-(3,4-Difluorophenyl)-3-(4-fluorophenyl)urea, 97%
Brand: Alfa Aesar™ H62653.03
Code : NEW
Additional Details : Weight : 0.00100kg
Quantity | 1g |
---|
Chemical Identifiers
1-(3,4-Difluorophenyl)-3-(4-fluorophenyl)urea | |
266.22 | |
MFCD07072794 |
Specifications
1-(3,4-Difluorophenyl)-3-(4-fluorophenyl)urea | |
Solid | |
266.22 |
1g | |
97% | |
MFCD07072794 |