missing translation for 'onlineSavingsMsg'
Learn More

1-[3-(4-chlorophenyl)-5-methylisoxazol-4-yl]ethan-1-one, 97%, Thermo Scientific™

Product Code. 10385202
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10385202 10 g 10g
10652083 1 g 1g
10311785 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10385202 Supplier Thermo Scientific Maybridge Supplier No. SPB04957EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 169814-48-6
Molecular Formula C12H10ClNO2
Molecular Weight (g/mol) 235.67
MDL Number MFCD01313507
InChI Key XSAMMAPYCPHHKH-UHFFFAOYSA-N
Synonym 1-3-4-chlorophenyl-5-methylisoxazol-4-yl ethan-1-one, 1-3-4-chlorophenyl-5-methylisoxazol-4-yl ethanone, 1-3-4-chlorophenyl-5-methyl-1,2-oxazol-4-yl ethanone, 3-4-chlorophenyl-4-acetyl-5-methylisoxazole, ethanone, 1-3-4-chlorophenyl-5-methyl-4-isoxazolyl, maybridge1_008689, 4-acetyl-3-4-chlorophenyl-5-methylisoxazole, 1-3-4-chlorophenyl-5-methyl-4-isoxazolyl-ethanone, 1-3-4-chloro-phenyl-5-methyl-isoxazol-4-yl-ethanone
PubChem CID 2735240
IUPAC Name 1-[3-(4-chlorophenyl)-5-methyl-1,2-oxazol-4-yl]ethan-1-one
SMILES CC(=O)C1=C(C)ON=C1C1=CC=C(Cl)C=C1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 235.67
Percent Purity 97%
Chemical Name or Material 1-[3-(4-Chlorophenyl)-5-methylisoxazol-4-yl]ethan-1-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.