missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3,4,6-Tetra-O-acetyl-2-O-trifluoromethanesulfonyl-Beta-D-mannopyranose, TRC
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Specifications
Specifications
| Chemical Name or Material | Tetra-O-acetyl-mannose triflate |
| CAS | 92051-23-5 |
| Molecular Formula | C15 H19 F3 O12 S |
| Quantity | 250 mg |
| Synonym | Tetra-O-acetyl-mannose triflate, 1,3,4,6-Tetra-O-acetyl-2-O-trifluoromethanesulfonyl-β-d-mannopyranose, 1,3,4,6-tetraacetate 2-(trifluoromethanesulfonate)-β-D-Mannopyranose, 1,3,4,6-Tetra-O-acetyl-2-(trifluoromethylsulfonyl)-β-D-mannopyranose, 1,3,4,6-Tetra-O-acetyl-2-O-trifluoromethanesulfonyl-β-D-mannopyranose, Mannose triflate |
| InChI Formula | InChI=1S/C15H19F3O12S/c1-6(19)25-5-10-11(26-7(2)20)12(27-8(3)21)13(14(29-10)28-9(4)22)30-31(23,24)15(16,17)18/h10-14H,5H2,1-4H3/t10-,11-,12+,13+,14-/m1/s1 |
| SMILES | CC(=O)OC[C@H]1O[C@@H](OC(=O)C)[C@@H](OS(=O)(=O)C(F)(F)F)[C@@H](OC(=O)C)[C@@H]1OC(=O)C |
| IUPAC Name | [(2R,3R,4S,5S,6S)-3,4,6-triacetyloxy-5-(trifluoromethylsulfonyloxy)oxan-2-yl]methyl acetate |
| Molecular Weight (g/mol) | 480.36 |
| Formula Weight | 480.0549 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction