missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(3,4,5-Trimethoxybenzyl)piperazine Dihydrochloride, Mikromol™
Description
1-(3,4,5-Trimethoxybenzyl)piperazine Dihydrochloride
Specifications
Specifications
| Chemical Name or Material | 1-(3,4,5-Trimethoxybenzyl)piperazine Dihydrochloride |
| CAS | 107049-57-0 |
| Analyte or Component Names | 1-(3,4,5-Trimethoxybenzyl)piperazine Dihydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C14 H22 N2 O3 . 2 Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | Trimetazidine Dihydrochloride Imp. A (EP), Trimetazidine Imp. A (EP), 1-(3,4,5-Trimethoxybenzyl)piperazine Dihydrochloride, Trimetazidine Dihydrochloride Imp. A (EP) as Dihydrochloride, Trimetazidine Dihydrochloride Impurity A as Dihydrochloride, Trimetazidine Impurity A as Dihydrochloride |
| InChI Formula | InChI=1S/C14H22N2O3.2ClH/c1-17-12-8-11(9-13(18-2)14(12)19-3)10-16-6-4-15-5-7-16;;/h8-9,15H,4-7,10H2,1-3H3;2*1H |
| SMILES | Cl.Cl.COc1cc(CN2CCNCC2)cc(OC)c1OC |
| IUPAC Name | 1-[(3,4,5-trimethoxyphenyl)methyl]piperazine;dihydrochloride |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?