missing translation for 'onlineSavingsMsg'
Learn More

1-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]pyrrolidine, 97%, Thermo Scientific™

Product Code. 10352983
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10352983 1 g Amber Glass Bottle 1g
11326795 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10352983 Supplier Thermo Scientific Maybridge Supplier No. CC55739DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 884507-45-3
Molecular Formula C17H26BNO2
Molecular Weight (g/mol) 287.21
MDL Number MFCD08741425
InChI Key AKQLICZDOCQKRA-UHFFFAOYSA-N
Synonym 1-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzyl pyrrolidine, 1-3-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl pyrrolidine, 1-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl pyrrolidine, amtb769, 3-pyrrolidinomethyl phenylboronic acid, pinacol ester, 3-pyrrolidine methyl phenylboronic acid pinacol ester, pyrrolidine,1-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl
PubChem CID 18525872
SMILES CC1(C)OB(OC1(C)C)C1=CC(CN2CCCC2)=CC=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
IUPAC Name 1-[[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl]pyrrolidine
Formula Weight 287.21
Percent Purity 97%
Chemical Name or Material 1-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]pyrrolidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.