missing translation for 'onlineSavingsMsg'
Learn More

1-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]piperidine, Thermo Scientific™

Product Code. 10585164
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 10585164

Brand: Thermo Scientific Maybridge CC45239DE

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 859833-21-9
Molecular Formula C18H28BNO2
Molecular Weight (g/mol) 301.24
MDL Number MFCD08435871
InChI Key KEKNXVTYDJCRMT-UHFFFAOYSA-N
Synonym 1-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzyl piperidine, 1-3-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl piperidine, 1-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl piperidine, piperidine,1-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl, 3-piperidine methyl phenylboronic acid pinacol ester, amtb771, 3-piperidin-1-ylmethyl phenylboronic acid, pinacol ester, 2-3-piperidinomethyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PubChem CID 16414184
IUPAC Name 1-[[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl]piperidine
SMILES CC1(C)OB(OC1(C)C)C1=CC(CN2CCCCC2)=CC=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 301.24
Percent Purity 97%
Chemical Name or Material 1-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]piperidine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.