missing translation for 'onlineSavingsMsg'
Learn More

1,3,3-Trimethyl-2-methyleneindoline, 95%

Product Code. 11459230
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11459230 250 g 250g
11449230 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11459230 Supplier Thermo Scientific Alfa Aesar Supplier No. A16741.30
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 118-12-7 | C12H15N | 173.259 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118-12-7
Molecular Formula C12H15N
Molecular Weight (g/mol) 173.259
MDL Number MFCD00005813
InChI Key ZTUKGBOUHWYFGC-UHFFFAOYSA-N
Synonym 1,3,3-trimethyl-2-methyleneindoline, fischer's base, fischer base, 1h-indole, 2,3-dihydro-1,3,3-trimethyl-2-methylene, fischer's methylene base, 2-methylene-1,3,3-trimethylindoline, fischers base, ccris 6608, indoline, 2-methylene-1,3,3-trimethyl, indoline, 1,3,3-trimethyl-2-methylene
PubChem CID 8351
IUPAC Name 1,3,3-trimethyl-2-methylideneindole
SMILES CC1(C(=C)N(C2=CC=CC=C21)C)C

Specifications

Melting Point -10°C
Density 0.979
Boiling Point 247°C to 248°C
Flash Point 101°C (213°F)
Refractive Index 1.578
Quantity 250 g
Beilstein 131162
Formula Weight 173.26
Percent Purity 95%
Chemical Name or Material 1,3,3-Trimethyl-2-methyleneindoline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.