missing translation for 'onlineSavingsMsg'
Learn More

1-(3,3-Dimethyl-1,2-dioxopentyl)-L-proline 3-(3-pyridinyl)propyl Ester, TRC

Product Code. 30381116
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30381116 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30381116 Supplier Toronto Research Chemicals Supplier No. TRCD47269010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-(3,3-Dimethyl-1,2-dioxopentyl)-L-proline 3-(3-pyridinyl)propyl Ester
CAS 186452-09-5
Molecular Formula C20 H28 N2 O4
Quantity 10 mg
Synonym GPI 1046
InChI Formula InChI=1S/C20H28N2O4/c1-4-20(2,3)17(23)18(24)22-12-6-10-16(22)19(25)26-13-7-9-15-8-5-11-21-14-15/h5,8,11,14,16H,4,6-7,9-10,12-13H2,1-3H3/t16-/m0/s1
SMILES CCC(C)(C)C(=O)C(=O)N1CCC[C@H]1C(=O)OCCCc2cccnc2
IUPAC Name 3-pyridin-3-ylpropyl (2S)-1-(3,3-dimethyl-2-oxopentanoyl)pyrrolidine-2-carboxylate
Molecular Weight (g/mol) 360.45
Formula Weight 360.2049
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.