missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(3-(2-(Dimethylamino)ethyl)-1-((5-((N-methylsulfamoyl)methyl)-1H-indol-3-yl)methyl)-1H-indol-5-yl)-N-methylmethanesulfonamide Formate Salt, TRC
Specifications
Specifications
| Chemical Name or Material | 1-(3-(2-(Dimethylamino)ethyl)-1-((5-((N-methylsulfamoyl)methyl)-1H-indol-3-yl)methyl)-1H-indol-5-yl)-N-methylmethanesulfonamide Formate Salt |
| Molecular Formula | C25 H33 N5 O4 S2 . C H2 O2 |
| Quantity | 5 mg |
| Synonym | Sumatriptan Impurity Formate Salt |
| InChI Formula | InChI=1S/C25H33N5O4S2.CH2O2/c1-26-35(31,32)16-18-5-7-24-22(11-18)21(13-28-24)15-30-14-20(9-10-29(3)4)23-12-19(6-8-25(23)30)17-36(33,34)27-2;2-1-3/h5-8,11-14,26-28H,9-10,15-17H2,1-4H3;1H,(H,2,3) |
| SMILES | O=S(CC1=CC(C(CN2C=C(CCN(C)C)C3=C2C=CC(CS(NC)(=O)=O)=C3)=CN4)=C4C=C1)(NC)=O.[H]C(O)=O |
| Molecular Weight (g/mol) | 531.694603 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?