missing translation for 'onlineSavingsMsg'
Learn More

1-[[(2RS)-2-(2,4-Difluorophenyl)oxiran-2-yl]methyl]-1H-1,2,4-triazole Mesilate, Mikromol™

Product Code. 30462412
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
This item is not returnable. View return policy

Product Code. 30462412

Brand: Mikromol MM0235.0800254AMP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

1-[[(2RS)-2-(2,4-Difluorophenyl)oxiran-2-yl]methyl]-1H-1,2,4-triazole Mesilate, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-[((2RS)-2-(2,4-Difluorophenyl)oxiran-2-yl)methyl]-1H-1,2,4-triazole Mesilate
CAS 86386-77-8
Analyte or Component Names 1-[((2RS)-2-(2,4-Difluorophenyl)oxiran-2-yl)methyl]-1H-1,2,4-triazole Mesilate
Shipping Condition Room Temperature
Molecular Formula C11 H9 F2 N3 O . C H4 O3 S
Certifications/Compliance ISO 17025
Synonym 1H-1,2,4-Triazole, 1-[[2-(2,4-difluorophenyl)-2-oxiranyl]methyl]-, methanesulfonate (1:1), 1H-1,2,4-Triazole, 1-[[2-(2,4-difluorophenyl)oxiranyl]methyl]-, monomethanesulfonate (9CI), 2-(2,4-Difluorophenyl)-2,3-epoxy-1-(1H-1,2,4-triazol-1-yl)propane methanesulfonate salt, 1-[[(2RS)-2-(2,4-Difluorophenyl)oxiran-2-yl]methyl]-1H-1,2,4-triazole mesilate, Fluconazole Impurity G as Mesilate
InChI Formula InChI=1S/C11H9F2N3O.CH4O3S/c12-8-1-2-9(10(13)3-8)11(5-17-11)4-16-7-14-6-15-16;1-5(2,3)4/h1-3,6-7H,4-5H2;1H3,(H,2,3,4)
Solution Type Neat
SMILES CS(=O)(=O)O.Fc1ccc(c(F)c1)C2(Cn3cncn3)CO2
IUPAC Name 1-[[2-(2,4-difluorophenyl)oxiran-2-yl]methyl]-1,2,4-triazole;methanesulfonic acid
Molecular Weight (g/mol) 333.31
Quantity 4 x 25 mg
Formula Weight 333.0595 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.