missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-[[(2RS)-2-(2,4-Difluorophenyl)oxiran-2-yl]methyl]-1H-1,2,4-triazole Mesilate, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
Description
1-[[(2RS)-2-(2,4-Difluorophenyl)oxiran-2-yl]methyl]-1H-1,2,4-triazole Mesilate
Specifications
Specifications
| Chemical Name or Material | 1-[((2RS)-2-(2,4-Difluorophenyl)oxiran-2-yl)methyl]-1H-1,2,4-triazole Mesilate |
| CAS | 86386-77-8 |
| Analyte or Component Names | 1-[((2RS)-2-(2,4-Difluorophenyl)oxiran-2-yl)methyl]-1H-1,2,4-triazole Mesilate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C11 H9 F2 N3 O . C H4 O3 S |
| Certifications/Compliance | ISO 17025 |
| Synonym | 1H-1,2,4-Triazole, 1-[[2-(2,4-difluorophenyl)-2-oxiranyl]methyl]-, methanesulfonate (1:1), 1H-1,2,4-Triazole, 1-[[2-(2,4-difluorophenyl)oxiranyl]methyl]-, monomethanesulfonate (9CI), 2-(2,4-Difluorophenyl)-2,3-epoxy-1-(1H-1,2,4-triazol-1-yl)propane methanesulfonate salt, 1-[[(2RS)-2-(2,4-Difluorophenyl)oxiran-2-yl]methyl]-1H-1,2,4-triazole mesilate, Fluconazole Impurity G as Mesilate |
| InChI Formula | InChI=1S/C11H9F2N3O.CH4O3S/c12-8-1-2-9(10(13)3-8)11(5-17-11)4-16-7-14-6-15-16;1-5(2,3)4/h1-3,6-7H,4-5H2;1H3,(H,2,3,4) |
| SMILES | CS(=O)(=O)O.Fc1ccc(c(F)c1)C2(Cn3cncn3)CO2 |
| IUPAC Name | 1-[[2-(2,4-difluorophenyl)oxiran-2-yl]methyl]-1,2,4-triazole;methanesulfonic acid |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction