missing translation for 'onlineSavingsMsg'
Learn More

1-[(2RS)-2-(2,4-Dichlorophenyl)-2-[[4-(benzenesulfonyl)benzyl]oxy]ethyl]-1H-imidazole Nitrate (Fenticonazole Sulfone Nitrate), Mikromol™

Product Code. 17903026
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17903026 25 mg 25mg
17942706 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17903026 Supplier Mikromol Supplier No. MM1216.050025
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

1-[(2RS)-2-(2,4-Dichlorophenyl)-2-[[4-(benzenesulfonyl)benzyl]oxy]ethyl]-1H-imidazole Nitrate (Fenticonazole Sulfone Nitrate)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fenticonazole Sulfone Nitrate
CAS 80676-29-5
Analyte or Component Names Fenticonazole Sulfone Nitrate
Shipping Condition Room Temperature
Molecular Formula C24 H20 Cl2 N2 O3 S . H N O3
Certifications/Compliance ISO 17025
Synonym 1-[2-(2,4-Dichlorophenyl)-2-[[4-(phenylsulfonyl)phenyl]methoxy]ethyl]-1H-imidazole Nitrate, Fenticonazole Sulfone Nitrate, 1-[(2RS)-2-(2,4-Dichlorophenyl)-2-[[4-(benzenesulfonyl)benzyl]oxy]ethyl]-1H-imidazole Nitrate, 1H-Imidazole, 1-[2-(2,4-dichlorophenyl)-2-[[4-(phenylsulfonyl)phenyl]methoxy]ethyl]-, nitrate (1:1), 1H-Imidazole, 1-[2-(2,4-dichlorophenyl)-2-[[4-(phenylsulfonyl)phenyl]methoxy]ethyl]-, mononitrate (9CI), Fenticonazole impurity C, Fenticonazole Nitrate Impurity C as Nitrate, Fenticonazole Impurity C as Nitrate
InChI Formula InChI=1S/C24H20Cl2N2O3S.HNO3/c25-19-8-11-22(23(26)14-19)24(15-28-13-12-27-17-28)31-16-18-6-9-21(10-7-18)32(29,30)20-4-2-1-3-5-20;2-1(3)4/h1-14,17,24H,15-16H2;(H,2,3,4)
SMILES ON(=O)=O.Clc1ccc(C(Cn2ccnc2)OCc3ccc(cc3)S(=O)(=O)c4ccccc4)c(Cl)c1
IUPAC Name 1-[2-[[4-(benzenesulfonyl)phenyl]methoxy]-2-(2,4-dichlorophenyl)ethyl]imidazole;nitrate
Molecular Weight (g/mol) 550.41
Quantity 25 mg
Formula Weight 549.0528 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.