missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trajan Scientific and Medical™ 1.2mm ID Direct Straight-through Liners
The unique deactivation process of SGE inlet liners provides excellent analysis for a wide range of compounds
Brand: Trajan Scientific and Medical™ 09201601
Code : 13
Additional Details : Weight : 0.10000kg
Description
- Each batch of liners tested for activity
- Manufactured from borosilicate glass
- Color coded by geometry for easy identification
- For use with Agilent and Thermo Scientific instruments
Specifications
GC Inlet Liner, Direct, Straight-Through Liner | |
78.5 mm | |
6.3 mm |
1 Ea. | |
1.2 mm | |
Agilent and Thermo Scientific Instruments |