missing translation for 'onlineSavingsMsg'
Learn More

Trajan Scientific and Medical™ 1.2mm ID Direct Straight-through Liners

The unique deactivation process of SGE inlet liners provides excellent analysis for a wide range of compounds

Brand:  Trajan Scientific and Medical™ 092016

Code : 13

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10350321

  • £84.90 / Pack of 5
Estimated Shipment: 29-05-2024
to see stock.
Add to basket

Description

Description

    Each batch of liners tested for activity
  • Manufactured from borosilicate glass
  • Color coded by geometry for easy identification
  • For use with Agilent and Thermo Scientific instruments
Specifications

Specifications

GC Inlet Liner, Direct, Straight-Through Liner
78.5 mm
6.3 mm
5/Pk.
1.2 mm
Agilent and Thermo Scientific Instruments
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers