missing translation for 'onlineSavingsMsg'
Learn More

1(2H)-Phthalazinone, 98+%

Product Code. 10390564 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10390564 25 g Glass bottle 25g
10541324 5 g Glass bottle 5g
10228373 100 g Glass bottle 100g
10483355 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10390564 Supplier Thermo Scientific Acros Supplier No. 384570250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 119-39-1 | C8H6N2O | 146.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119-39-1
Molecular Formula C8H6N2O
Molecular Weight (g/mol) 146.15
MDL Number MFCD00006892
InChI Key IJAPPYDYQCXOEF-UHFFFAOYSA-N
Synonym 1 2h-phthalazinone, phthalazin-1 2h-one, phthalazone, phthalazinone, phthalazin-1-one, 1-phthalazinol, phthalazin-1-ol, 1-hydroxyphthalazine, 1 2h phthalazinone, 1,2-dihydrophthalazin-1-one
PubChem CID 8394
ChEBI CHEBI:34023
IUPAC Name 2H-phthalazin-1-one
SMILES C1=CC=C2C(=C1)C=NNC2=O

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 184.0°C to 187.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Formula Weight 146.15
Percent Purity 98+%
Physical Form Crystalline Powder
Chemical Name or Material 1(2H)-Phthalazinone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.