missing translation for 'onlineSavingsMsg'
Learn More

1,24-Diethyl Tetracosanedioate, TRC

Product Code. 30369347
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30369347 50 mg 50mg
30381454 100 mg -
30374578 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30369347 Supplier Toronto Research Chemicals Supplier No. TRCD53172550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1472-91-9
Molecular Formula C28H54O4
Molecular Weight (g/mol) 454.73
Synonym Tetracosanedioic Acid Diethyl Ester
IUPAC Name diethyl tetracosanedioate
SMILES CCOC(=O)CCCCCCCCCCCCCCCCCCCCCCC(=O)OCC

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C28H54O4/c1-3-31-27(29)25-23-21-19-17-15-13-11-9-7-5-6-8-10-12-14-16-18-20-22-24-26-28(30)32-4-2/h3-26H2,1-2H3
Formula Weight 454.4022
Chemical Name or Material 1,24-Diethyl Tetracosanedioate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.