missing translation for 'onlineSavingsMsg'
Learn More

1-[2-(Trifluoromethyl)phenyl]hydrazine, ≥95%, Maybridge

Product Code. 10212362
Change view
Click to view available options
:
Amber Glass Bottle; 1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10212362 Amber Glass Bottle; 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10212362 Supplier Thermo Scientific Maybridge Supplier No. RF01370DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 365-34-4
CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Molecular Formula C7H7F3N2
Linear Formula CF3C6H4NHNH2
MDL Number MFCD00052687
Quantity 1 g
Synonym 2-trifluoromethyl phenylhydrazine, 2-trifluoromethyl phenyl hydrazine, 2-trifluoromethylphenylhydrazine, 1-2-trifluoromethyl phenyl hydrazine, m-trifluoromethylphenylhydrazine, hydrazine, 2-trifluoromethyl phenyl, buttpark 44\57-17, 2-trifluoromethyl-phenyl-hydrazine
InChI Key JSWQDLBFVSTSIW-UHFFFAOYSA-N
SMILES C1=CC=C(C(=C1)C(F)(F)F)NN
IUPAC Name [2-(trifluoromethyl)phenyl]hydrazine
Molecular Weight (g/mol) 176.142
PubChem CID 520800
Formula Weight 176.14
Percent Purity 95%
Chemical Name or Material 1-[2-(Trifluoromethyl)phenyl]hydrazine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.