missing translation for 'onlineSavingsMsg'
Learn More

1-[2-(Trifluoromethyl)phenyl]-2-propanone, TRC

Product Code. 30454950
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30454950 250 mg 250mg
30451486 100 mg 100mg
30449251 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30454950 Supplier Toronto Research Chemicals Supplier No. TRCT774545250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21235-67-6
Molecular Formula C10H9F3O
Molecular Weight (g/mol) 202.17
Synonym 1-[2-(Trifluoromethyl)phenyl]-2-propanone, 1-(alpha,alpha,alpha-Trifluoro-o-tolyl)-2-propanone
IUPAC Name 1-[2-(trifluoromethyl)phenyl]propan-2-one
SMILES CC(=O)Cc1ccccc1C(F)(F)F

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C10H9F3O/c1-7(14)6-8-4-2-3-5-9(8)10(11,12)13/h2-5H,6H2,1H3
Formula Weight 202.0605
Chemical Name or Material 1-[2-(Trifluoromethyl)phenyl]-2-propanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.