missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Tetrahydropyranyl)-1H-pyrazole, 98%, Thermo Scientific™

Product Code. 15481349
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15481349

Brand: Thermo Scientific Alfa Aesar H64246.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 449758-17-2
Molecular Formula C8H12N2O
Molecular Weight (g/mol) 152.197
MDL Number MFCD11044692
InChI Key IMZWSOSYNFVECD-UHFFFAOYSA-N
Synonym 1-2-tetrahydropyranyl-1h-pyrazole, 1-tetrahydro-2h-pyran-2-yl-1h-pyrazole, 1-oxan-2-yl-1h-pyrazole, 1-oxan-2-yl pyrazole, 1h-pyrazole, 1-tetrahydro-2h-pyran-2-yl, 1-2-tetrahydropyranyl pyrazole, 1-tetrahydro-pyran-2-yl-1h-pyrazole, 1-2-oxanyl pyrazole, ksc588k5l, 1-tetrahydropyran-2-ylpyrazole
PubChem CID 21866310
IUPAC Name 1-(oxan-2-yl)pyrazole
SMILES C1CCOC(C1)N2C=CC=N2

Specifications

Color Colorless to Yellow
Density 1.084 g/mL
Boiling Point 60°C to 65°C (0.8 mmHg)
Flash Point 104°C (219°F)
Refractive Index 1.503
Quantity 1 g
Beilstein 9694988
Formula Weight 152.2
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-(2-Tetrahydropyranyl)-1H-pyrazole
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.