missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Pyrimidinyl)piperazine dihydrochloride, 98+%, Thermo Scientific™

Product Code. 11441087
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11441087 100 g 100g
11421087 5 g 5g
11431087 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11441087 Supplier Thermo Scientific Alfa Aesar Supplier No. B24085.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94021-22-4
Molecular Formula C8H14Cl2N4
Molecular Weight (g/mol) 237.128
MDL Number MFCD00012779
InChI Key ZNZGJSLHXOMREP-UHFFFAOYSA-N
Synonym 2-1-piperazinyl pyrimidine dihydrochloride, 1-2-pyrimidyl piperazine dihydrochloride, 2-piperazin-1-yl pyrimidine dihydrochloride, 1-2-pyrimidinyl piperazine dihydrochloride, 2-piperazinylpyrimidine hcl, 2-piperazinylpyrimidine dihydrochloride, 2-piperazin-1-ylpyrimidine dihydrochloride, pyrimidine, 2-1-piperazinyl-, dihydrochloride, pyrimidinylpiperazine dihydrochloride, pubchem7349
PubChem CID 11680272
IUPAC Name 2-piperazin-1-ylpyrimidine;dihydrochloride
SMILES C1CN(CCN1)C2=NC=CC=N2.Cl.Cl

Specifications

Melting Point ∼280°C (decomposition)
Quantity 100 g
Beilstein 151178
Sensitivity Hygroscopic
Formula Weight 237.13
Percent Purity ≥98%
Chemical Name or Material 1-(2-Pyrimidinyl)piperazine dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.