missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Pyridylazo)-2-Naphthol, Pure, Fisher Chemical™

Product Code. 10598060 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Jar
Vial
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10598060 1 g Vial 1g
10050310 5 g Glass Jar 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10598060 Supplier Fisher Chemical Supplier No. P/8000/43

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. Vea la política de devoluciones

Identificadores de productos químicos

CAS 85-85-8
Molecular Formula C15H11N3O
Molecular Weight (g/mol) 249.273
MDL Number 4071
InChI Key RAXUMGMWXZYADR-OBGWFSINSA-N
Synonym 1-2-pyridylazo-2-naphthol, unii-095b53y3xv, pan indicator, 2-naphthalenol, 1-2-pyridinylazo, 1e-1-pyridin-2-ylhydrazinylidene naphthalen-2-one, 1-2-pyridylazo-2-hydroxynaphthalene, pan van, 1-2-pyridyldiazenyl naphthalen-2-ol, 1-pyridin-2-azo-2-naphthol, 2-naphthalenol, 1-2-2-pyridinyl diazenyl
PubChem CID 5376264
IUPAC Name (1E)-1-(pyridin-2-ylhydrazinylidene)naphthalen-2-one
SMILES C1=CC=C2C(=C1)C=CC(=O)C2=NNC3=CC=CC=N3

Especificaciones

Melting Point 138°C
Color Red
Packaging Vial
Quantity 1 g
Formula Weight 249.27g/mol
Physical Form Powder
compliance-icons
Nombre del producto
  • 1-(2-Pyridylazo)-2-naphthol
Palabra de advertencia
  • Warning
Clasificación
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Indicaciones de peligro
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Consejos de prudencia
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.