missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(2-Pyridylazo)-2-naphthol, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
1-(2-Pyridylazo)-2-naphthol is widely used as an indicator for complexometry. It is used as a reagent for the extraction and spectrophotometric determination (for colorimetric analysis) of many transition metal elements like arsenic, chromium, cobalt, copper, cadmium, manganese, zinc and zirconium. It is also used for the analysis of uranium and thorium in solutions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-(2-Pyridylazo)-2-naphthol is widely used as an indicator for complexometry. It is used as a reagent for the extraction and spectrophotometric determination (for colorimetric analysis) of many transition metal elements like arsenic, chromium, cobalt, copper, cadmium, manganese, zinc and zirconium. It is also used for the analysis of uranium and thorium in solutions.
Solubility
Insoluble in water.
Notes
Incompatible with strong bases and strong oxidizing agents.
1-(2-Pyridylazo)-2-naphthol is widely used as an indicator for complexometry. It is used as a reagent for the extraction and spectrophotometric determination (for colorimetric analysis) of many transition metal elements like arsenic, chromium, cobalt, copper, cadmium, manganese, zinc and zirconium. It is also used for the analysis of uranium and thorium in solutions.
Solubility
Insoluble in water.
Notes
Incompatible with strong bases and strong oxidizing agents.
Specifications
Melting Point | 138°C to 141°C |
Quantity | 5 g |
Beilstein | 206390 |
Solubility Information | Insoluble in water. |
Formula Weight | 249.27 |
Percent Purity | 98% |
Chemical Name or Material | 1-(2-Pyridylazo)-2-naphthol |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'