missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Pyridin-3-ylphenyl)methanamine dihydrochloride, ≥97%, Thermo Scientific™

Product Code. 10085872
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10085872 1 g Amber Glass Bottle 1g
10758633 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10085872 Supplier Thermo Scientific Maybridge Supplier No. CC41863DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 859833-18-4
Molecular Formula C12H14Cl2N2
Molecular Weight (g/mol) 257.158
MDL Number MFCD08060476
InChI Key SSFBEGXFELCUJT-UHFFFAOYSA-N
Synonym 1-2-pyridin-3-ylphenyl methanamine dihydrochloride, 1-2-pyridin-3-ylphenyl methylamine dihydrochloride, 2-pyridin-3-yl phenyl methanamine dihydrochloride, 1-2-pyridin-3-ylphenyl methanaminedihydrochloride, 1-2-pyridin-3-yl phenyl methanamine dihydrochloride, 2-pyridin-3-yl-benzylamine dihydrochloride, 2-3-pyridinyl benzenemethanamine dihydrochloride, 2-3-pyridyl phenyl methylamine, chloride, chloride
PubChem CID 18525754
IUPAC Name (2-pyridin-3-ylphenyl)methanamine;dihydrochloride
SMILES C1=CC=C(C(=C1)CN)C2=CN=CC=C2.Cl.Cl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 257.16
Percent Purity 97%
Chemical Name or Material 1-(2-Pyridin-3-ylphenyl)methanamine dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.