missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-1,2-Propylene-d6 Carbonate, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | 1,2-Propylene-d6 Carbonate |
| CAS | 202480-74-8 |
| Molecular Formula | C4 2H6 O3 |
| Quantity | 1 g |
| InChI Formula | InChI=1 S/C4H6O3/c1-3-2-6-4(5)7-3/h3H,2H2,1H3/i1D3,2D2,3 D |
| SMILES | [2 H]C([2 H])([2 H])C1([2 H])OC(=O)OC1([2 H])[2 H] |
| IUPAC Name | 4,4,5-trideuterio-5-(trideuteriomethyl)-1,3-dioxolan-2-one |
| Molecular Weight (g/mol) | 108.13 |
| Formula Weight | 108.0694 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction