missing translation for 'onlineSavingsMsg'
Learn More

1,2-Propanediol monomethyl ether acetate, 99%, stab. with 50ppm BHT

Product Code. p-7046633
Click to view available options
Quantity:
100 mL
1000 mL
2500 mL
Unit Size:
1 L
100mL
2.5 L
This item is not returnable. View return policy

Product Code. 11368266

Brand: Thermo Scientific Alfa Aesar L15459.AE

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 108-65-6 | C6H12O3 | 132.159 g/mol

1,2-Propanediol monomethyl ether acetate is used as a solvent for paints, inks, lacquers, varnishes, cleaners and coatings. It is also used as a solvent for degreasing circuit boards and in food contact applications. Further, it is used in photoresist formulations in the semiconductor industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Propanediol monomethyl ether acetate is used as a solvent for paints, inks, lacquers, varnishes, cleaners and coatings. It is also used as a solvent for degreasing circuit boards and in food contact applications. Further, it is used in photoresist formulations in the semiconductor industry.

Solubility
Miscible with water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong acids, strong bases, reducing agents, acids and alkalis.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-65-6
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.159
MDL Number MFCD00038500
InChI Key LLHKCFNBLRBOGN-UHFFFAOYSA-N
Synonym 1-methoxy-2-propyl acetate, pgmea, 2-acetoxy-1-methoxypropane, propylene glycol monomethyl ether acetate, propylene glycol methyl ether acetate, 1-methoxy-2-acetoxypropane, 2-propanol, 1-methoxy-, acetate, 2-methoxy-1-methylethyl acetate, propyleneglycol monomethyl ether acetate, acetic acid, 2-methoxy-1-methylethyl ester
PubChem CID 7946
IUPAC Name 1-methoxypropan-2-yl acetate
SMILES CC(COC)OC(=O)C

Specifications

Melting Point -67°C
Density 0.968
pH 4
Boiling Point 145°C to 146°C
Flash Point 43°C (109°F)
Odor Ester-like
Refractive Index 1.402
Quantity 100 mL
UN Number UN3272
Beilstein 1751656
Solubility Information Miscible with water.
Formula Weight 132.16
Percent Purity 99%
Chemical Name or Material 1,2-Propanediol monomethyl ether acetate, stabilized with 50ppm BHT
Show More Show Less
compliance-icons
Product Identifier
  • 1,2-Propanediol monomethyl ether acetate
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
  • MIXTURE LIST-Contains : 1,2-Propanediol monomethyl ether acetate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.