missing translation for 'onlineSavingsMsg'
Learn More

1,2-Propanediol, 98+%

Product Code. 11468337
Change view
Click to view available options
Quantity:
500 g
2500 g
Unit Size:
2500g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11468337 2500 g 2500g
11458337 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11468337 Supplier Thermo Scientific Alfa Aesar Supplier No. A18406.0E
  • ONLINE EXCLUSIVE PRICE
    £62.93
    £51.65 / 2500g
    Save £11.28
    17% Off
View all promotions
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 57-55-6 | C3H8O2 | 76.095 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-55-6
Molecular Formula C3H8O2
Molecular Weight (g/mol) 76.095
MDL Number MFCD00064272
InChI Key DNIAPMSPPWPWGF-UHFFFAOYSA-N
Synonym 1,2-propanediol, propylene glycol, 1,2-dihydroxypropane, 2-hydroxypropanol, 1,2-propylene glycol, methylethyl glycol, methylethylene glycol, monopropylene glycol, isopropylene glycol, dowfrost
PubChem CID 1030
ChEBI CHEBI:16997
IUPAC Name propane-1,2-diol
SMILES CC(CO)O

Specifications

Melting Point -60°C
Density 1.036
pH Neutral
Boiling Point 186°C to 188°C
Flash Point 107°C (225°F)
Odor Nearly Odorless
Refractive Index 1.432
Quantity 2500 g
Beilstein 1340498
Sensitivity Hygroscopic
Merck Index 14,7855
Solubility Information Miscible with water,benzene,acetone,chloroform and ether. Immiscible with fixed oils.
Formula Weight 76.1
Percent Purity ≥98%
Chemical Name or Material 1,2-Propanediol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.